Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028855.1 | NP_083131.1 | 3 | 795 | ||
GenBank mRNA | AK007080.1 | - | 3 | 811 | ||
BC048760.1 | - | 3 | 795 | |||
BC100505.1 | - | 3 | 787 | 30917608 | ||
BC144838.1 | - | 1 | 1159 | |||
EF120626.1 | - | 3 | 795 |