Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270988.1 | NP_001257917.1 | 12 | 2991 | ||
NM_172421.5 | NP_766009.2 | 11 | 2847 | |||
GenBank mRNA | AK036839.1 | - | 11 | 2804 | ||
AK122540.1 | - | 3 | 1620 | |||
AK162455.1 | - | 3 | 1588 | |||
BC141140.1 | - | 12 | 2825 | 9055752 |