Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 4, 5 | 413 | ||
GenBank mRNA | AB047550.1 | - | 4, 5 | 425 | ||
AB076609.1 | - | 3, 4 | 304 | |||
AK016312.1 | - | 4, 5 | 430 | |||
AK043091.1 | - | 4, 5 | 414 | |||
AK049559.1 | - | 4, 5 | 406 | |||
AK050796.1 | - | 3, 4 | 282 | |||
AK077443.1 | - | 4, 5 | 413 | |||
AK159468.1 | - | 4, 5 | 428 | |||
AK167173.1 | - | 4, 5 | 421 | |||
AK167202.1 | - | 4, 5 | 421 | |||
AX775963.1 | - | 4, 5 | 383 | |||
BC004013.1 | - | 4, 5 | 407 | 3493738 |