Search
Search

Fam187b| Gene Symbol: Fam187b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242647.1 | NP_001229576.1 | 2 | 941 | ||
| NM_175240.4 | NP_780449.2 | 2 | 969 | |||
| GenBank mRNA | AK006143.1 | - | 2 | 966 | ||
| AK028081.1 | - | 2 | 970 | |||
| AK148492.1 | - | 2 | 704 | |||
| BC019737.1 | - | 2 | 683 | |||
| BC120864.1 | - | 2 | 970 | 40129787 | ||
| BC125560.1 | - | 2 | 970 | 40130075 | ||