Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CYP20A1 SIRNA Assays ›
Gene Symbol:
Cyp20a1
Gene Name:
cytochrome P450, family 20, subfamily a, polypeptide 1
Gene Aliases:
A930011N14Rik, Cypm
Chromosome Location:
Chr.1: 60342800 - 60388060 on Build GRCm38
UniGene ID:
Mm.197640
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS234126
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cyp20a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_030013.2
NP_084289.1
10
1017
GenBank mRNA
AK053388.1
-
10
1020
BC049147.1
-
9
834
6306072
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidoreductase
oxygenase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
oxidation-reduction process
monooxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
fluorene oxygenase activity
mono-butyltin dioxygenase activity
tri-n-butyltin dioxygenase activity
di-n-butyltin dioxygenase activity
methylsilanetriol hydroxylase activity
methyl tertiary butyl ether 3-monooxygenase activity
4-nitrocatechol 4-monooxygenase activity
4-chlorophenoxyacetate monooxygenase activity
tert-butanol 2-monooxygenase activity
alpha-pinene monooxygenase activity
dimethylsilanediol hydroxylase activity
ammonia monooxygenase activity
hydroxymethylsilanetriol oxidase activity
2-hydroxyisobutyrate 3-monooxygenase activity
alpha-pinene dehydrogenase activity
heme binding
bisphenol A hydroxylase B activity
2,2-bis(4-hydroxyphenyl)-1-propanol hydroxylase activity
9-fluorenone-3,4-dioxygenase activity
anthracene 9,10-dioxygenase activity
2-(methylthio)benzothiazole monooxygenase activity
2-hydroxybenzothiazole monooxygenase activity
benzothiazole monooxygenase activity
2,6-dihydroxybenzothiazole monooxygenase activity
pinacolone 5-monooxygenase activity
thioacetamide S-oxygenase activity
thioacetamide S-oxide S-oxygenase activity
endosulfan monooxygenase I activity
N-nitrodimethylamine hydroxylase activity
4-(1-ethyl-1,4-dimethyl-pentyl)phenol monoxygenase activity
endosulfan ether monooxygenase activity
pyrene 4,5-monooxygenase activity
pyrene 1,2-monooxygenase activity
1-hydroxypyrene 6,7-monooxygenase activity
1-hydroxypyrene 7,8-monooxygenase activity
phenylboronic acid monooxygenase activity
spheroidene monooxygenase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0