Search
Search

Chia1| Gene Symbol: Chia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023186.3 | NP_075675.2 | 6 | 512 | ||
| GenBank mRNA | AF154571.1 | - | 6 | 506 | ||
| AF290003.1 | - | 6 | 511 | |||
| AK007573.1 | - | 6 | 524 | |||
| AK008633.1 | - | 6 | 525 | |||
| AK008650.2 | - | 6 | 525 | |||
| AK008757.1 | - | 6 | 525 | |||
| AK160173.1 | - | 6 | 525 | |||
| BC011134.1 | - | 6 | 507 | 4165150 | ||
| BC034548.1 | - | 6 | 512 | 4189080 | ||
| DQ349202.1 | - | 6 | 509 | |||
| EF094027.1 | - | 6 | 509 | |||