Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL23A SIRNA Assays ›
Gene Symbol:
Il23a
Gene Name:
interleukin 23, alpha subunit p19
Gene Aliases:
IL-23, p19
Chromosome Location:
Chr.10: 128296140 - 128298084 on Build GRCm38
UniGene ID:
Mm.125482
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS234680
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il23a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031252.2
NP_112542.1
3
488
GenBank mRNA
AF301619.1
-
3
488
AK043894.1
-
1375
BC019953.1
-
3
472
3708687
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
interleukin superfamily
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
immune system process
positive regulation of T-helper 1 type immune response
inflammatory response
immune response
positive regulation of activation of JAK2 kinase activity
negative regulation of interleukin-10 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell proliferation
T cell proliferation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat4 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
positive regulation of memory T cell differentiation
innate immune response
positive regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
tissue remodeling
defense response to Gram-negative bacterium
positive regulation of NK T cell activation
positive regulation of NK T cell proliferation
defense response to virus
positive regulation of neutrophil chemotaxis
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 17 cell lineage commitment
cytokine activity
protein binding
interleukin-23 receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0