Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요?
계정 생성하기
모든 제품 주문
항체
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR 분석
피펫 및 피펫 팁
실험실용 원심분리기
초저온 냉동고
분광학
비이커
PCR 장비 및 용품
제품 카테고리 전체보기
응용 분야 및 기법
세포 분석
실험실 장비
Real-Time PCR
PCR
크로마토그래피
세포 배양 및 트랜스펙션
DNA/RNA 추출과 분석
단백질 생물학
유세포분석
화학 제품
응용 분야 및 기법 전체보기
서비스
맞춤형 서비스
엔터프라이즈급 실험실 정보 처리
엔터프라이즈 서비스
360° CDMO 및 CRO 서비스
CDMO 서비스
임상 시험 CRO 서비
장비 서비스
교육 서비스
서비스 전체 보기
지원
고객센터
문의하기
시험성적서(COA) 및 적합성 인증서(COC)
Safety Data Sheets (SDS)
매뉴얼
인용 및 참고 문헌
기기 지원
기술 자료/제품 FAQ
학습 센터
지원 메뉴 전체보기
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
문의하기
빠른 주문
주문 현황
제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
전체검색
Search button
주문 현황
빠른주문
로그인
로그인
회원이 아니신가요?
계정 생성하기
계정정보
주문내역조회
커스텀제품 및 프로젝트
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL23A SIRNA Assays ›
Gene Symbol:
Il23a
Gene Name:
interleukin 23, alpha subunit p19
Gene Aliases:
IL-23, p19
Chromosome Location:
Chr.10: 128296140 - 128298084 on Build GRCm38
UniGene ID:
Mm.125482
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS234681
Size
20 nmol
20 nmol
재고 정보
주문접수 후 제작
Catalog #
1320001
제품 정가
Contact Us
Your Price
온라인 행사가:
공급가 확인 ›
카트에 추가
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il23a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031252.2
NP_112542.1
2, 3
374
GenBank mRNA
AF301619.1
-
2, 3
374
AK043894.1
-
1261
BC019953.1
-
2, 3
358
3708687
Back To Top
추가 정보
Panther Classification:
Molecular Function -
cytokine
interleukin superfamily
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
immune system process
positive regulation of T-helper 1 type immune response
inflammatory response
immune response
positive regulation of activation of JAK2 kinase activity
negative regulation of interleukin-10 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell proliferation
T cell proliferation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat4 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
positive regulation of memory T cell differentiation
innate immune response
positive regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
tissue remodeling
defense response to Gram-negative bacterium
positive regulation of NK T cell activation
positive regulation of NK T cell proliferation
defense response to virus
positive regulation of neutrophil chemotaxis
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 17 cell lineage commitment
cytokine activity
protein binding
interleukin-23 receptor binding
Back To Top
관련 제품
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0