Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133871.2 | NP_598632.2 | 7 | 1243 | ||
GenBank mRNA | AJ299405.1 | - | 6 | 1133 | ||
AK079749.1 | - | 7 | 1195 | |||
AK136553.1 | - | 7 | 1195 | |||
AK151346.1 | - | 7 | 1194 | |||
AK171676.1 | - | 7 | 1176 | |||
BC026901.1 | - | 2 | 141 | |||
BC111882.1 | - | 6 | 1133 | 40060245 | ||
BC112402.1 | - | 6 | 1133 |