Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DXO SIRNA Assays ›
Gene Symbol:
Dxo
Gene Name:
decapping exoribonuclease
Gene Aliases:
Dom3z, NG6
Chromosome Location:
Chr.17: 34837019 - 34839186 on Build GRCm38
UniGene ID:
Mm.275309
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS235743
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dxo
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163770.1
NP_001157242.1
3
759
NM_033613.2
NP_291091.2
3
784
GenBank mRNA
BC004713.1
-
3
720
3499698
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_212497, BC082083.
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
metabolic process
RNA destabilization
nuclear mRNA surveillance
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
mRNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
thiamine-pyrophosphatase activity
5'-3' exonuclease activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
RNA pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ckh64:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0