Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AXIN2 SIRNA Assays ›
          Gene Symbol:
          Axin2
          Gene Name:
          axin2
          Gene Aliases:
          Axi1, Axil, Conductin
          Chromosome Location:
          Chr.11: 108917828 - 108950783 on Build GRCm38
          UniGene ID:
          Mm.71710
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS236017
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Axin2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015732.4 NP_056547.3 10 2632
          GenBank mRNA AF073788.1 - 10 2497
          AF205889.1 - 9 2337
          AK049192.1 - 10 2576
          AK084644.1 - 3 417
          AK142333.1 - 10 2634
          AK202759.1 - 2 153
          BC057338.1 - 10 2615 6827741

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          somitogenesis
          positive regulation of protein phosphorylation
          intramembranous ossification
          secondary heart field specification
          chondrocyte differentiation involved in endochondral bone morphogenesis
          regulation of transcription, DNA-templated
          cell death
          cell proliferation
          negative regulation of cell proliferation
          dorsal/ventral axis specification
          positive regulation of epithelial to mesenchymal transition
          positive regulation of cell death
          Wnt signaling pathway
          regulation of Wnt signaling pathway
          bone mineralization
          regulation of mismatch repair
          cellular protein localization
          regulation of catenin import into nucleus
          negative regulation of catenin import into nucleus
          termination of G-protein coupled receptor signaling pathway
          odontogenesis
          positive regulation of GTPase activity
          maintenance of DNA repeat elements
          negative regulation of osteoblast differentiation
          positive regulation of protein kinase activity
          mRNA stabilization
          regulation of chondrocyte development
          regulation of centromeric sister chromatid cohesion
          cellular response to organic cyclic compound
          negative regulation of canonical Wnt signaling pathway
          Wnt signaling pathway involved in somitogenesis
          negative regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          GTPase activator activity
          protein binding
          beta-catenin binding
          enzyme binding
          protein kinase binding
          ubiquitin protein ligase binding
          armadillo repeat domain binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0