Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L11 SIRNA Assays ›
          Gene Symbol:
          Bcl2l11
          Gene Name:
          BCL2-like 11 (apoptosis facilitator)
          Gene Aliases:
          1500006F24Rik, Bim, Bod, bcl2-L-11
          Chromosome Location:
          Chr.2: 128126038 - 128162547 on Build GRCm38
          UniGene ID:
          Mm.141083
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS236049
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bcl2l11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009754.3 NP_033884.1 3, 4 465
          NM_207680.2 NP_997563.1 3, 4 723
          NM_207681.2 NP_997564.1 4, 5 555
          GenBank mRNA AF032459.1 - 2, 3 495
          AF032460.1 - 3, 4 327
          AF032461.1 - 2, 3 237
          AK011490.1 - 3, 4 712
          AK050192.1 - 4, 5 544
          AK082613.1 - 3, 4 719
          AK150464.1 - 3, 4 466
          AK152472.1 - 3, 4 466
          AK153542.1 - 3, 4 466
          BC019556.1 - 3, 4 717 4459720

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          leukocyte homeostasis
          B cell homeostasis
          B cell apoptotic process
          kidney development
          lymphocyte homeostasis
          myeloid cell homeostasis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell-matrix adhesion
          spermatogenesis
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          post-embryonic development
          positive regulation of cell death
          mammary gland development
          positive regulation of protein homooligomerization
          response to endoplasmic reticulum stress
          tube formation
          odontogenesis of dentin-containing tooth
          regulation of apoptotic process
          T cell homeostasis
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of neuron apoptotic process
          ear development
          positive regulation of cell cycle
          regulation of organ growth
          developmental pigmentation
          regulation of developmental pigmentation
          spleen development
          thymus development
          post-embryonic organ morphogenesis
          positive regulation of apoptotic process by virus
          cellular process regulating host cell cycle in response to virus
          thymocyte apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of mitochondrial membrane permeability involved in apoptotic process
          apoptotic process involved in embryonic digit morphogenesis
          positive regulation of fibroblast apoptotic process
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          microtubule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4lrsz:80/100.66.79.169:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0