Search
Search

Il11| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290423.1 | NP_001277352.1 | 4, 5 | 477 | ||
| NM_008350.4 | NP_032376.1 | 4, 5 | 576 | |||
| GenBank mRNA | AK132897.1 | - | 4, 5 | 478 | ||
| BC134354.1 | - | 4, 5 | 438 | 40054168 | ||
| U03421.1 | - | 4, 5 | 469 | |||