Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL1B SIRNA Assays ›
          Gene Symbol:
          Il1b
          Gene Name:
          interleukin 1 beta
          Gene Aliases:
          IL-1beta, Il-1b
          Chromosome Location:
          Chr.2: 129363843 - 129371173 on Build GRCm38
          UniGene ID:
          Mm.222830
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS236889
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il1b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008361.3 NP_032387.1 5 511
          GenBank mRNA AK156396.1 - 4 1063
          AK157245.1 - 4 1063
          AK168047.1 - 5 531
          AK224980.1 - 4 1063
          AK225002.1 - 5 530
          AK225003.1 - 1 86
          BC011437.1 - 5 511 3989461
          M15131.1 - 5 520

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          activation of MAPK activity
          fever generation
          positive regulation of protein phosphorylation
          inflammatory response
          immune response
          positive regulation of cytosolic calcium ion concentration
          aging
          memory
          negative regulation of cell proliferation
          response to carbohydrate
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of glucose transport
          positive regulation of cell death
          negative regulation of glutamate secretion
          cytokine-mediated signaling pathway
          hyaluronan biosynthetic process
          neutrophil chemotaxis
          sequestering of triglyceride
          positive regulation of fever generation
          lipopolysaccharide-mediated signaling pathway
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          interleukin-1 beta production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of stress-activated MAPK cascade
          positive regulation of immature T cell proliferation in thymus
          response to ATP
          positive regulation of heterotypic cell-cell adhesion
          social behavior
          ectopic germ cell programmed cell death
          positive regulation of myosin light chain kinase activity
          cellular response to drug
          positive regulation of T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAP kinase activity
          protein kinase B signaling
          positive regulation of JUN kinase activity
          positive regulation of chemokine biosynthetic process
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of neuron differentiation
          positive regulation of glial cell differentiation
          positive regulation of angiogenesis
          negative regulation of lipid metabolic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          negative regulation of insulin receptor signaling pathway
          positive regulation of astrocyte differentiation
          positive regulation of phagocytosis
          negative regulation of neurogenesis
          regulation of insulin secretion
          leukocyte migration
          negative regulation of lipid catabolic process
          positive regulation of lipid catabolic process
          positive regulation of membrane protein ectodomain proteolysis
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of cell division
          positive regulation of calcidiol 1-monooxygenase activity
          negative regulation of adiponectin secretion
          positive regulation of ERK1 and ERK2 cascade
          monocyte aggregation
          cellular response to organic substance
          cellular response to organic cyclic compound
          positive regulation of monocyte chemotactic protein-1 production
          positive regulation of neutrophil chemotaxis
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of branching morphogenesis of a nerve
          negative regulation of neural precursor cell proliferation
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          receptor binding
          cytokine activity
          interleukin-1 receptor binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0