Search
Search

Lmx1b| Gene Symbol: Lmx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010725.2 | NP_034855.2 | 2, 3 | 249 | ||
| GenBank mRNA | AB221651.1 | - | 2, 3 | 249 | ||
| AF078166.1 | - | 2, 3 | 249 | |||
| BC119169.1 | - | 2, 3 | 328 | 8733918 | ||
| BC125469.1 | - | 2, 3 | 328 | 40129984 | ||