Search
Search

Rab8a| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023126.2 | NP_075615.2 | 1 | 173 | ||
| GenBank mRNA | AB232600.1 | - | 1 | 14 | ||
| AK076048.1 | - | 1 | 52 | |||
| AK079306.1 | - | 40 | ||||
| AK080740.1 | - | 1 | 25 | |||
| AK147178.1 | - | 1 | 41 | |||
| AK165690.1 | - | 1 | 36 | |||
| BC019990.1 | - | 1 | 14 | 4161110 | ||
| S53270.1 | - | 1 | 31 | |||