Search
Search

Bspry| Gene Symbol: Bspry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138653.1 | NP_619594.1 | 6 | 1035 | ||
| GenBank mRNA | AJ276690.1 | - | 6 | 1035 | ||
| AK035588.1 | - | 6 | 1041 | |||
| AK160930.1 | - | 6 | 1041 | |||
| BC003992.1 | - | 4 | 578 | |||
| BC139079.1 | - | 6 | 1051 | 8862101 | ||
| BC139080.1 | - | 6 | 1051 | 8862102 | ||