Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037764.1 | NP_001032853.1 | 2 | 1255 | ||
NM_009021.2 | NP_033047.2 | 3 | 1478 | |||
GenBank mRNA | AK154252.1 | - | 3 | 1317 | ||
AY548172.1 | - | 2 | 1258 | |||
BC145882.1 | - | 1 | 978 | 40131185 | ||
BC145884.1 | - | 1 | 978 | 40131187 | ||
D29801.1 | - | 2 | 1258 |