Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RARA SIRNA Assays ›
          Gene Symbol:
          Rara
          Gene Name:
          retinoic acid receptor, alpha
          Gene Aliases:
          Nr1b1, RAR, RARalpha1
          Chromosome Location:
          Chr.11: 98927809 - 98974942 on Build GRCm38
          UniGene ID:
          Mm.439744
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS237735
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rara
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001176528.1 NP_001169999.1 3 884
          NM_001177302.1 NP_001170773.1 5 1089
          NM_001177303.1 NP_001170774.1 4 834
          NM_009024.2 NP_033050.2 4 956
          GenBank mRNA AB221572.1 - 3 403
          AK042163.1 - 4 835
          AK153644.1 - 3 430
          AK154623.1 - 4 929
          AK170956.1 - 3 884
          BC010216.1 - 4 954 3498142
          BC038266.1 - 3 953
          BC040383.1 - 5 1089
          BC090396.1 - 3 500
          CT010231.1 - 3 399
          S56656.1 - 4 936

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001145301, NM_001024809, NM_000964, AK292205, AK098172, and others.
          This siRNA also targets the following transcripts in Rat: NM_031528, XM_008767945, AJ002941, AJ002940, BC099830, and others.

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          neural tube closure
          glandular epithelial cell development
          outflow tract septum morphogenesis
          growth plate cartilage development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein phosphorylation
          signal transduction
          germ cell development
          spermatogenesis
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of translation
          regulation of granulocyte differentiation
          negative regulation of granulocyte differentiation
          embryonic camera-type eye development
          regulation of myelination
          response to retinoic acid
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-5 production
          multicellular organism growth
          regulation of apoptotic process
          negative regulation of apoptotic process
          apoptotic cell clearance
          steroid hormone mediated signaling pathway
          regulation of phosphatidylinositol 3-kinase activity
          response to ethanol
          negative regulation of cell differentiation
          positive regulation of T-helper 2 cell differentiation
          positive regulation of neuron differentiation
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of translational initiation
          regulation of synaptic plasticity
          retinoic acid receptor signaling pathway
          positive regulation of binding
          positive regulation of protein kinase B signaling
          ventricular cardiac muscle cell differentiation
          Sertoli cell fate commitment
          limb development
          face development
          bone development
          trachea cartilage development
          chondroblast differentiation
          negative regulation of cartilage development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to estrogen stimulus
          translation repressor activity, nucleic acid binding
          regulatory region DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region DNA binding
          retinoic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          transcription coactivator activity
          transcription corepressor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          receptor binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          enzyme binding
          protein domain specific binding
          chromatin DNA binding
          phosphatidylinositol 3-kinase regulator activity
          histone deacetylase binding
          protein kinase B binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          retinoic acid-responsive element binding
          metal ion binding
          protein heterodimerization activity
          mRNA 5'-UTR binding
          protein kinase A binding
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zl7kt:80/100.66.73.1:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0