Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025285.2 | NP_079561.1 | 5 | 598 | ||
GenBank mRNA | AK003659.1 | - | 5 | 598 | ||
AK013020.1 | - | 5 | 598 | |||
AK149212.1 | - | 5 | 623 | |||
BC026538.1 | - | 5 | 560 | 4981703 |