Search
Search

Mageb18| Gene Symbol: Mageb18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173783.3 | NP_776144.1 | 5 | 956 | ||
| GenBank mRNA | AK046641.1 | - | 5 | 958 | ||
| BC116801.1 | - | 2 | 684 | 40126120 | ||
| BC137691.1 | - | 2 | 684 | 8860710 | ||