Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146045.1 | NP_666157.1 | 5 | 781 | ||
GenBank mRNA | AK141739.1 | - | 5 | 745 | ||
AK146855.1 | - | 5 | 828 | |||
AK171600.1 | - | 5 | 797 | |||
BC027195.1 | - | 5 | 781 | 4224150 |