Search
Search

Lrfn4| Gene Symbol: Lrfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153388.4 | NP_700437.2 | 2 | 2010 | ||
| GenBank mRNA | AK081560.1 | - | 1 | 2293 | ||
| AK163603.1 | - | 2 | 1868 | |||
| BC023036.1 | - | 1 | 1623 | 5358628 | ||
| BC023156.1 | - | 2 | 1666 | 4951386 | ||
| DQ078786.1 | - | 1 | 1295 | |||