Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YAP1 SIRNA Assays ›
          Gene Symbol:
          Yap1
          Gene Name:
          yes-associated protein 1
          Gene Aliases:
          AI325207, Yap, Yap65, Yki, Yorkie
          Chromosome Location:
          Chr.9: 7931999 - 8004596 on Build GRCm38
          UniGene ID:
          Mm.221992
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS238824
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Yap1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171147.1 NP_001164618.1 5 986
          NM_009534.3 NP_033560.1 5 986
          GenBank mRNA AK182200.1 - 3 170
          AK204312.1 - 1 24
          BC014733.1 - 5 983 4239820
          BC039125.1 - 5 841 2649070
          BC094313.1 - 5 974 5697882
          X80508.1 - 5 978

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001034002, XM_006242489, XM_006242490, XM_008765894, XM_006242492, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          vasculogenesis
          embryonic heart tube morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          cell proliferation
          positive regulation of cell proliferation
          regulation of keratinocyte proliferation
          keratinocyte differentiation
          negative regulation of epithelial cell differentiation
          notochord development
          somatic stem cell population maintenance
          hippo signaling
          regulation of cell proliferation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of organ growth
          paraxial mesoderm development
          lateral mesoderm development
          regulation of neurogenesis
          contact inhibition
          bud elongation involved in lung branching
          lung epithelial cell differentiation
          regulation of canonical Wnt signaling pathway
          cellular response to retinoic acid
          cellular response to gamma radiation
          regulation of stem cell proliferation
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of canonical Wnt signaling pathway
          positive regulation of stem cell population maintenance
          negative regulation of nucleic acid-templated transcription
          negative regulation of stem cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          protein C-terminus binding
          transcription regulatory region DNA binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0