Search
Search

Zbtb17| Gene Symbol: Zbtb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009541.2 | NP_033567.2 | 13 | 1998 | ||
| GenBank mRNA | AK051971.1 | - | 12 | 2069 | ||
| AK134251.1 | - | 13 | 1958 | |||
| BC060055.1 | - | 10 | 2268 | |||
| BC145312.1 | - | 12 | 1871 | 9053867 | ||
| BC145313.1 | - | 11 | 1998 | |||
| BC145821.1 | - | 12 | 1871 | 40131124 | ||
| U14556.1 | - | 13 | 1996 | |||
| U22396.1 | - | 13 | 1986 | |||