Search
Search

Ficd| Gene Symbol: Ficd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010825.3 | NP_001010825.2 | 3 | 1387 | ||
| GenBank mRNA | AK044446.1 | - | 3 | 1196 | ||
| AK054325.1 | - | 3 | 1152 | |||
| AK087477.1 | - | 3 | 1371 | |||
| BC039952.1 | - | 1 | 621 | |||
| BC106090.1 | - | 3 | 1358 | |||
| BC139821.1 | - | 2 | 1024 | 40141450 | ||