Search
Search

Trappc1| Gene Symbol: Trappc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024206.2 | NP_001019377.1 | 4 | 284 | ||
| GenBank mRNA | BC115818.1 | - | 3 | 253 | 40107010 | |
| BC115921.1 | - | 3 | 253 | 40107011 | ||
| BY479576.1 | - | 2 | 145 | |||
| CK621128.1 | - | 4 | 284 | |||