Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD5 SIRNA Assays ›
Gene Symbol:
Chd5
Gene Name:
chromodomain helicase DNA binding protein 5
Gene Aliases:
4930532L22Rik, AW060752, B230399N07Rik, CHD-5
Chromosome Location:
Chr.4: 152338651 - 152390194 on Build GRCm38
UniGene ID:
Mm.40192
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS241047
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Chd5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081376.1
NP_001074845.1
15, 16
2746
NM_029216.2
NP_083492.2
15, 16
2746
GenBank mRNA
AK147563.1
-
15, 16
2747
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
nervous system development
negative regulation of cell proliferation
chromatin modification
cerebral cortex neuron differentiation
cell differentiation
spermatogenesis, exchange of chromosomal proteins
histone H4 acetylation
regulation of cell differentiation
regulation of transcription involved in cell fate commitment
histone H3-K27 trimethylation
positive regulation of signal transduction by p53 class mediator
nucleotide binding
nucleic acid binding
DNA binding
chromatin binding
helicase activity
protein binding
ATP binding
ATP-dependent helicase activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
histone deacetylase binding
metal ion binding
H3K27me3 modified histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qm47m:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0