Search
Search

Galntl6| Gene Symbol: Galntl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175032.3 | NP_778197.2 | 4 | 638 | ||
| GenBank mRNA | AK080614.1 | - | 4 | 638 | ||
| BB641428.1 | - | 4 | 638 | |||
| BC111102.1 | - | 5 | 681 | |||
| GU220061.1 | - | 4 | 485 | |||