Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trappc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013718.2 | NP_038746.1 | 4 | 457 | ||
GenBank mRNA | AF041433.1 | - | 4 | 448 | ||
AK151747.1 | - | 4 | 469 | |||
AK168518.1 | - | 4 | 497 | |||
BC003736.1 | - | 4 | 457 | 3490979 |