Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam135b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177819.3 | NP_808487.2 | 4 | 466 | ||
GenBank mRNA | AK043571.1 | - | 4 | 467 | ||
BC125645.1 | - | 4 | 341 | |||
BC125647.1 | - | 4 | 341 |