Search
Search

Nell1| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037906.2 | NP_001032995.1 | 4, 5 | 538 | ||
| GenBank mRNA | AK032845.1 | - | 4, 5 | 820 | ||
| AY622226.1 | - | 4, 5 | 538 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK035921.1 | - | 1027 | Chr. - - [-] on Build | ||