Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dnaic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034878.2 | NP_001030050.2 | 3 | 594 | ||
GenBank mRNA | AK138639.1 | - | 3 | 521 | ||
EF565270.1 | - | 2 | 210 |