Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019493.3 | NP_062366.2 | 5 | 951 | ||
GenBank mRNA | AB050983.2 | - | 5 | 951 | ||
AJ005120.1 | - | 5 | 824 | |||
AK135849.1 | - | 2 | 330 | |||
AK136041.1 | - | 5 | 938 | |||
AK136124.1 | - | 5 | 939 | |||
AK136149.1 | - | 5 | 941 | |||
AK136185.1 | - | 5 | 944 | |||
AK161379.1 | - | 7 | 1030 | |||
AK163407.1 | - | 5 | 939 |