Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291205.1 | NP_001278134.1 | 6 | 690 | ||
NM_001291206.1 | NP_001278135.1 | 6 | 1041 | |||
NM_001291207.1 | NP_001278136.1 | 5 | 586 | |||
NM_001291208.1 | NP_001278137.1 | 5 | 935 | |||
NM_001291210.1 | NP_001278139.1 | 4 | 464 | |||
NM_026446.4 | NP_080722.1 | 6 | 692 | |||
GenBank mRNA | AF535869.1 | - | 6 | 1042 | ||
AF535870.1 | - | 5 | 936 | |||
AK003967.1 | - | 2 | 108 | |||
AK011740.2 | - | 5 | 722 | |||
AK019401.1 | - | 6 | 665 | |||
AK042531.1 | - | 6 | 640 | |||
AK154844.1 | - | 6 | 692 | |||
AY138505.1 | - | 5 | 538 | |||
BC003838.1 | - | 6 | 624 | 3490927 |