Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019731.1 | NP_062705.1 | 3 | 294 | ||
GenBank mRNA | AF153451.1 | - | 3 | 294 | ||
AF288690.1 | - | 3 | 307 | |||
AK011573.1 | - | 2 | 475 | |||
AK012805.1 | - | 3 | 330 | |||
AK017724.1 | - | 3 | 310 | |||
BC027277.1 | - | 3 | 312 | 3591383 |