Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sult5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020564.3 | NP_065589.2 | 5 | 637 | ||
GenBank mRNA | AF026074.1 | - | 5 | 625 | ||
BC012677.1 | - | 5 | 633 | 4187578 | ||
CO044978.1 | - | 3 | 537 |