Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024919.1 | NP_001020090.1 | 3 | 669 | ||
GenBank mRNA | AK009485.1 | - | 3 | 651 | ||
AK208742.1 | - | 1 | 48 | |||
BC116435.1 | - | 4 | 501 | 40104481 | ||
BC116436.1 | - | 4 | 501 | 40104482 | ||
BC128508.1 | - | 4 | 571 | |||
BC128509.1 | - | 4 | 501 | 40104485 |