Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024919.1 | NP_001020090.1 | 3 | 573 | ||
GenBank mRNA | AK009485.1 | - | 3 | 555 | ||
AK219559.1 | - | 2 | 224 | |||
BB616782.1 | - | 3 | 596 |