Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 7 | 879 | ||
NM_001190886.1 | NP_001177815.1 | 6 | 835 | |||
NM_001290690.1 | NP_001277619.1 | 6 | 1034 | |||
NM_027398.3 | NP_081674.2 | 6 | 846 | |||
GenBank mRNA | AB075041.1 | - | 6 | 476 | ||
AJ278534.1 | - | 5 | 404 | |||
AK014313.1 | - | 6 | 835 | |||
AK045948.1 | - | 7 | 880 | |||
AK046398.1 | - | 7 | 868 | |||
AK081845.1 | - | 6 | 836 | |||
AY050525.1 | - | 7 | 484 | |||
AY050526.1 | - | 6 | 451 | |||
AY171234.1 | - | 6 | 910 | |||
AY647242.1 | - | 6 | 678 | |||
BC034241.1 | - | 6 | 788 | 4503251 | ||
BU702946.1 | - | 5 | 532 | |||
DQ148493.1 | - | 6 | 788 | |||
DQ148494.1 | - | 6 | 836 | |||
DQ148495.1 | - | 7 | 484 |