Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027649.3 | NP_081925.2 | 5, 6 | 721 | ||
GenBank mRNA | AK005865.1 | - | 5, 6 | 722 | ||
AY336501.1 | - | 5, 6 | 609 | |||
BC131909.1 | - | 5, 6 | 681 | 40130186 |