Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2410016O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1088 | ||
GenBank mRNA | AB041553.1 | - | 1 | 1027 | ||
AK010521.1 | - | 1 | 194 | |||
AK080870.1 | - | 1 | 1034 | |||
BC052152.2 | - | 1 | 1002 | |||
BC059809.1 | - | 1 | 1039 | |||
BC138094.1 | - | 1 | 1088 | 8861113 |