Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 12 | 1959 | ||
GenBank mRNA | AK011892.1 | - | 8 | 890 | ||
AK046669.1 | - | 12 | 1925 | |||
AK144950.1 | - | 3032 | ||||
AK173311.1 | - | 10 | 3917 | |||
BC031392.1 | - | 12 | 1903 | 4952407 |