Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 12 | 1994 | ||
GenBank mRNA | AK011892.1 | - | 8 | 925 | ||
AK046669.1 | - | 12 | 1960 | |||
AK173311.1 | - | 10 | 3952 | |||
BC031392.1 | - | 12 | 1938 | 4952407 |