Search
Search

Dbr1| Gene Symbol: Dbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031403.3 | NP_113580.2 | 2 | 416 | ||
| GenBank mRNA | AK050090.1 | - | 2 | 417 | ||
| AK090351.1 | - | 2 | 392 | |||
| AK139631.1 | - | 2 | 398 | |||
| BC006661.1 | - | 2 | 411 | 3501390 | ||