Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134100.4 | NP_598861.1 | 2 | 1284 | ||
GenBank mRNA | AK087550.1 | - | 2 | 1289 | ||
AK089268.1 | - | 2 | 1287 | |||
AK165774.1 | - | 2 | 1259 | |||
BC009140.1 | - | 1 | 1191 | 3598007 |