Search
Search

Nucb1| Gene Symbol: Nucb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163662.1 | NP_001157134.1 | 9 | 955 | ||
| NM_008749.2 | NP_032775.1 | 9 | 955 | |||
| GenBank mRNA | AK002630.1 | - | 9 | 936 | ||
| AK004886.1 | - | 9 | 926 | |||
| AK007976.1 | - | 9 | 929 | |||
| AK045087.1 | - | 9 | 955 | |||
| AK140710.1 | - | 9 | 933 | |||
| AK160981.1 | - | 9 | 942 | |||
| AK164088.1 | - | 9 | 937 | |||
| AK170717.1 | - | 9 | 950 | |||
| AK194015.1 | - | 4 | 223 | |||
| AK209002.1 | - | 3 | 161 | |||
| BC072554.1 | - | 9 | 933 | 5684979 | ||
| M96823.1 | - | 8 | 877 | |||