Search
Search

Spi1| Gene Symbol: Spi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011355.1 | NP_035485.1 | 1, 2 | 205 | ||
| GenBank mRNA | AK142246.1 | - | 1, 2 | 222 | ||
| AK153524.1 | - | 1, 2 | 236 | |||
| AK154945.1 | - | 1, 2 | 257 | |||
| AK155085.1 | - | 1, 2 | 241 | |||
| BC003815.1 | - | 1, 2 | 123 | 3600260 | ||
| L03215.1 | - | 1, 2 | 102 | |||
| M32370.1 | - | 1, 2 | 185 | |||
| M38252.1 | - | 1, 2 | 205 | |||
| X17463.1 | - | 1, 2 | 893 | |||