Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Six2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011380.2 | NP_035510.1 | 1 | 566 | ||
GenBank mRNA | BC068021.1 | - | 1 | 566 | 6394139 | |
D83147.1 | - | 1 | 559 | |||
X80338.1 | - | 1 | 339 |