Search
Search

Prune| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173347.2 | NP_775482.1 | 7 | 1301 | ||
| GenBank mRNA | AF051908.2 | - | 7 | 848 | ||
| AK081637.1 | - | 7 | 992 | |||
| AK142160.1 | - | 7 | 900 | |||
| AK218289.1 | - | 2 | 177 | |||
| BC044877.1 | - | 5 | 726 | |||
| BC057546.1 | - | 7 | 1301 | 6416282 | ||
| BC058635.1 | - | 7 | 1030 | 5698271 | ||